Learn More
Hach Company Hach SC4500 Controller, Prognosys, mA Analog Outputs - Configured for 2 Analog UPW Conductivity Sensors, 100-240 VAC, without power cord (MPN LXV525.99A11661)
Catalog No. 5000207366 Shop All Hach Company Products
Encompass
The SC4500 is a digital process controller configured with Prognosys predictive diagnostics and mA analog outputs for ultrapure water conductivity monitoring. This configuration supports two analog UPW conductivity sensors, operates on 100-240 VAC, and ships without a power cord. It provides flexible analog and digital communications for integration into process control and remote monitoring systems.
  • Modern touchscreen interface for straightforward operator interaction and configuration.
  • Built-in predictive diagnostics to reduce unplanned downtime.
  • Multiple analog and digital connectivity options for flexible system integration.
  • Supports two analog UPW conductivity sensors for ultrapure water monitoring.
  • Expandable I/O with analog modules and optional digital communication protocols.
  • LAN, Wi-Fi, and cellular network options for remote access and data transfer.
  • Compact ½ DIN form factor for easy panel mounting.
Catalog No. 50-002-07366 Supplier Hach Company Supplier No. LXV525.99A11661
The SC4500 is a digital process controller configured with Prognosys predictive diagnostics and mA analog outputs for ultrapure water conductivity monitoring. This configuration supports two analog UPW conductivity sensors, operates on 100-240 VAC, and ships without a power cord. It provides flexible analog and digital communications for integration into process control and remote monitoring systems.
  • Modern touchscreen interface for straightforward operator interaction and configuration.
  • Built-in predictive diagnostics to reduce unplanned downtime.
  • Multiple analog and digital connectivity options for flexible system integration.
  • Supports two analog UPW conductivity sensors for ultrapure water monitoring.
  • Expandable I/O with analog modules and optional digital communication protocols.
  • LAN, Wi-Fi, and cellular network options for remote access and data transfer.
  • Compact ½ DIN form factor for easy panel mounting.