Learn More
Hach Company Hach SC4x00 USB Stick, MPN LXZ525.99.00017 - Accessory for SC4x00 Controllers (SC4500, SC1500)
Catalog No. 5000207526 Shop All Hach Company Products
Encompass
The SC4x00 USB Stick (MPN LXZ525.99.00017) is a removable storage accessory from Hach for SC4x00 series controllers. It provides a simple way to transfer configurations, back up data, and apply firmware updates for compatible controllers. The manufacturer lists availability, price, and documentation on the product page.
  • Provides removable storage for configurations and instrument data.
  • Enables easy transfer of settings between controllers.
  • Supports firmware updates and backup of critical files.
  • Uses a plug-and-play USB interface for straightforward operation.
  • Includes access to product manuals and technical resources.
Catalog No. 50-002-07526 Supplier Hach Company Supplier No. LXZ525.99.00017
The SC4x00 USB Stick (MPN LXZ525.99.00017) is a removable storage accessory from Hach for SC4x00 series controllers. It provides a simple way to transfer configurations, back up data, and apply firmware updates for compatible controllers. The manufacturer lists availability, price, and documentation on the product page.
  • Provides removable storage for configurations and instrument data.
  • Enables easy transfer of settings between controllers.
  • Supports firmware updates and backup of critical files.
  • Uses a plug-and-play USB interface for straightforward operation.
  • Includes access to product manuals and technical resources.