Learn More
Hach Company Hach Sension+ 5202 pH Liquid-Filled Combination Electrode with Screw Cap S7 (MPN LZW5202.97.0002)
Catalog No. 5000207291 Shop All Hach Company Products
Encompass
The Hach Sension+ 5202 is a liquid-filled, refillable pH combination electrode designed for general laboratory and aqueous monitoring applications. It provides fast responding pH measurement from a large semi-spheric glass membrane and a ceramic junction, with a robust glass body and an S7 screw-cap connector for straightforward installation and replacement.
  • Refillable liquid electrolyte for stable reference potential.
  • Large semi-spheric glass membrane for fast response.
  • Ceramic junction suitable for challenging media.
  • Glass body for chemical resistance in aqueous environments.
  • S7 screw-cap connector for easy mounting and replacement.
  • Measuring range 0-12 pH and operating range 0-80 °C.
  • Designed for municipal and seawater monitoring applications.
Catalog No. 50-002-07291 Supplier Hach Company Supplier No. LZW5202.97.0002
The Hach Sension+ 5202 is a liquid-filled, refillable pH combination electrode designed for general laboratory and aqueous monitoring applications. It provides fast responding pH measurement from a large semi-spheric glass membrane and a ceramic junction, with a robust glass body and an S7 screw-cap connector for straightforward installation and replacement.
  • Refillable liquid electrolyte for stable reference potential.
  • Large semi-spheric glass membrane for fast response.
  • Ceramic junction suitable for challenging media.
  • Glass body for chemical resistance in aqueous environments.
  • S7 screw-cap connector for easy mounting and replacement.
  • Measuring range 0-12 pH and operating range 0-80 °C.
  • Designed for municipal and seawater monitoring applications.