Learn More
Hach Company Hach Sension+ pH Combination Electrode 5012T with A.T.C., Glass Body, 1 m Cable
Catalog No. 5000207288 Shop All Hach Company Products
Encompass
The Hach Sension+ pH combination electrode 5012T is a laboratory-grade electrode designed for reliable pH measurement in general aqueous samples. It includes automatic temperature compensation via an integrated Pt1000 sensor and a stable liquid reference for consistent results across a range of temperatures and sample types.
  • Automatic temperature compensation with built-in Pt1000 sensor.
  • Measuring range 0-14 pH.
  • Operating temperature 0-80 °C.
  • 1 m cable suitable for benchtop meters.
  • Liquid electrolyte (KCl 3M AgCl saturated) for stable reference.
  • Ceramic diaphragm for reliable junction performance.
  • Glass body designed for laboratory use.
  • Suitable for general aqueous, municipal, and sea water samples.
Catalog No. 50-002-07288 Supplier Hach Company Supplier No. LZW5012T.97.002
The Hach Sension+ pH combination electrode 5012T is a laboratory-grade electrode designed for reliable pH measurement in general aqueous samples. It includes automatic temperature compensation via an integrated Pt1000 sensor and a stable liquid reference for consistent results across a range of temperatures and sample types.
  • Automatic temperature compensation with built-in Pt1000 sensor.
  • Measuring range 0-14 pH.
  • Operating temperature 0-80 °C.
  • 1 m cable suitable for benchtop meters.
  • Liquid electrolyte (KCl 3M AgCl saturated) for stable reference.
  • Ceramic diaphragm for reliable junction performance.
  • Glass body designed for laboratory use.
  • Suitable for general aqueous, municipal, and sea water samples.