Learn More
Aqua Solutions For high range silica determination by the Silicomolybdate method (Hach method 8185).
SDP
Catalog No. NC0951752
Encompass
For high range silica determination by the Silicomolybdate method (Hach method 8185). Range: up to 100 mg/L SiO2. Sample Size: 25 mL. 100 tests
Catalog No. NC0951752 Supplier Aqua Solutions Supplier No. HA2244300
For high range silica determination by the Silicomolybdate method (Hach method 8185). Range: up to 100 mg/L SiO2. Sample Size: 25 mL. 100 tests