Learn More
Hach Company Hach Small Bore Electrodeless (Inductive) Convertible Conductivity Sensor, CL, 50 ft Cable (3725E2T50)
Catalog No. 5000207531 Shop All Hach Company Products
Encompass
The Hach 3725E2T50 small-bore electrodeless (inductive) conductivity sensor delivers wide-range, low-maintenance conductivity measurement for process and water-treatment applications. It features a polypropylene body, a built-in Pt1000 RTD for temperature compensation, and a 15 m (50 ft) analog cable with a convertible mounting style for immersion, insertion, union, and sanitary installations.
  • Wide measuring range from 200 μS/cm to 2,000,000 μS/cm.
  • Inductive design eliminates electrode polarization and coating.
  • Built-in Pt1000 RTD provides temperature compensation.
  • Convertible mounting supports immersion, insertion, union, and sanitary installations.
  • Polypropylene body offers chemical resistance for demanding environments.
  • Includes 15 m (50 ft) cable and user manual for straightforward installation.
Catalog No. 50-002-07531 Supplier Hach Company Supplier No. 3725E2T50
The Hach 3725E2T50 small-bore electrodeless (inductive) conductivity sensor delivers wide-range, low-maintenance conductivity measurement for process and water-treatment applications. It features a polypropylene body, a built-in Pt1000 RTD for temperature compensation, and a 15 m (50 ft) analog cable with a convertible mounting style for immersion, insertion, union, and sanitary installations.
  • Wide measuring range from 200 μS/cm to 2,000,000 μS/cm.
  • Inductive design eliminates electrode polarization and coating.
  • Built-in Pt1000 RTD provides temperature compensation.
  • Convertible mounting supports immersion, insertion, union, and sanitary installations.
  • Polypropylene body offers chemical resistance for demanding environments.
  • Includes 15 m (50 ft) cable and user manual for straightforward installation.