Learn More
Hach Company Hach Solitax ts-line sc Immersion Probe - Turbidity 0.001-4000 NTU; Suspended solids 0.001 mg/L-50 g/L; stainless steel; automatic wiper
Catalog No. NC3848508 Shop All Hach Company Products
Encompass
The Hach Solitax ts-line sc immersion probe provides continuous measurement of turbidity and total suspended solids (TSS) across an extended range for drinking water, wastewater, and industrial process monitoring. Stainless-steel construction and an automatic wiper support reliable operation in clear to highly turbid liquids and sludges, with simple integration into digital monitoring systems.
  • Measure turbidity from 0.001 to 4000 NTU.
  • Measure suspended solids from 0.001 mg/L to 50 g/L.
  • Immersion probe design for inline or immersion mounting.
  • Stainless-steel construction with corrosion-resistant optics carrier and sleeve.
  • Automatic wiper for sustained, low-maintenance operation.
  • Standard 10 m (33 ft) cable; extension cables available.
  • Sensor diameter approximately 60 mm and length approximately 200 mm.
  • Typical weight about 1.38 kg.
Catalog No. NC3848508 Supplier Hach Company Supplier No. LXV423.99.00100
Add to Cart
Add to Cart
Request a Quote
The Hach Solitax ts-line sc immersion probe provides continuous measurement of turbidity and total suspended solids (TSS) across an extended range for drinking water, wastewater, and industrial process monitoring. Stainless-steel construction and an automatic wiper support reliable operation in clear to highly turbid liquids and sludges, with simple integration into digital monitoring systems.
  • Measure turbidity from 0.001 to 4000 NTU.
  • Measure suspended solids from 0.001 mg/L to 50 g/L.
  • Immersion probe design for inline or immersion mounting.
  • Stainless-steel construction with corrosion-resistant optics carrier and sleeve.
  • Automatic wiper for sustained, low-maintenance operation.
  • Standard 10 m (33 ft) cable; extension cables available.
  • Sensor diameter approximately 60 mm and length approximately 200 mm.
  • Typical weight about 1.38 kg.