Learn More
Hach Company Hach SP510 Hardness Analyzer, 50 mg/L Trip Point
Catalog No. 5000207990 Shop All Hach Company Products
Encompass
The SP510 Hardness Analyzer is an inline water hardness monitor that continuously detects hardness breakthrough and triggers regeneration when hardness exceeds a preset 50 mg/L trip point, helping protect downstream equipment and optimize softener operation.
  • Continuously monitors total hardness and signals when the trip point is exceeded.
  • Preconfigured 50 mg/L trip point for automated regeneration control.
  • Colorimetric measurement using an LED with a 610 nm peak wavelength.
  • Includes reagents and installation and maintenance kits for startup and routine service.
  • Operates on 115/230 VAC, 70 VA, 50/60 Hz power supply.
  • ABS enclosure with IP62 rating for protected installations.
  • Recommended operating temperature range of 5-40 °C.
  • Simple maintenance: replenish reagents approximately every 2 months and replace pump tubing about every 6 months.
Catalog No. 50-002-07990 Supplier Hach Company Supplier No. 5410050
Add to Cart
Add to Cart
Request a Quote
The SP510 Hardness Analyzer is an inline water hardness monitor that continuously detects hardness breakthrough and triggers regeneration when hardness exceeds a preset 50 mg/L trip point, helping protect downstream equipment and optimize softener operation.
  • Continuously monitors total hardness and signals when the trip point is exceeded.
  • Preconfigured 50 mg/L trip point for automated regeneration control.
  • Colorimetric measurement using an LED with a 610 nm peak wavelength.
  • Includes reagents and installation and maintenance kits for startup and routine service.
  • Operates on 115/230 VAC, 70 VA, 50/60 Hz power supply.
  • ABS enclosure with IP62 rating for protected installations.
  • Recommended operating temperature range of 5-40 °C.
  • Simple maintenance: replenish reagents approximately every 2 months and replace pump tubing about every 6 months.