Learn More
Hach Company Hach TU5400sc Ultra-High Precision Low-Range Laser Turbidimeter with Automatic Cleaning, EPA version
Catalog No. 5000207391 Shop All Hach Company Products
Encompass
The TU5400sc is an ultra-high-precision low-range laser turbidimeter designed for sensitive, repeatable turbidity monitoring in regulatory and high-purity water applications. The EPA-approved TU5 platform combines a stable laser source and a 360° x 90° optical detection design with optional automatic cleaning to reduce maintenance and improve measurement reliability.
  • Provides measurement sensitivity down to 0.2 mNTU.
  • Uses 360° x 90° optical detection with a stable laser light source.
  • Offers an optional automatic cleaning module that reduces required maintenance.
  • Reduces measurement surface area by 98% to minimize cleaning time.
  • Designed for regulatory compliance and system check workflows.
  • Suitable for high-purity water, desalination, surface water, and industrial applications.
Catalog No. 50-002-07391 Supplier Hach Company Supplier No. LXV445.99.30212
Add to Cart
Add to Cart
Request a Quote
The TU5400sc is an ultra-high-precision low-range laser turbidimeter designed for sensitive, repeatable turbidity monitoring in regulatory and high-purity water applications. The EPA-approved TU5 platform combines a stable laser source and a 360° x 90° optical detection design with optional automatic cleaning to reduce maintenance and improve measurement reliability.
  • Provides measurement sensitivity down to 0.2 mNTU.
  • Uses 360° x 90° optical detection with a stable laser light source.
  • Offers an optional automatic cleaning module that reduces required maintenance.
  • Reduces measurement surface area by 98% to minimize cleaning time.
  • Designed for regulatory compliance and system check workflows.
  • Suitable for high-purity water, desalination, surface water, and industrial applications.