Learn More
Hach Company Hach Vial Compartment Wiper, Pack of 10 - For TU5300sc and TU5400sc
Catalog No. NC3797043 Shop All Hach Company Products
Encompass
The Hach vial compartment wiper (LZY910) is a pack of 10 cleaning wipers designed to maintain the vial compartment of TU5 series turbidimeters. Use these wipers to remove residue from the vial compartment interior, protecting internal optics and helping preserve measurement accuracy.
  • Pack of 10 cleaning wipers.
  • Designed to clean vial compartment interiors and protect optics.
  • Helps maintain measurement accuracy and reduce contamination.
  • Suitable for routine maintenance and calibration procedures.
  • Easy to use and replace as part of regular maintenance.
Catalog No. NC3797043 Supplier Hach Company Supplier No. LZY910
The Hach vial compartment wiper (LZY910) is a pack of 10 cleaning wipers designed to maintain the vial compartment of TU5 series turbidimeters. Use these wipers to remove residue from the vial compartment interior, protecting internal optics and helping preserve measurement accuracy.
  • Pack of 10 cleaning wipers.
  • Designed to clean vial compartment interiors and protect optics.
  • Helps maintain measurement accuracy and reduce contamination.
  • Suitable for routine maintenance and calibration procedures.
  • Easy to use and replace as part of regular maintenance.