Learn More
Hach Company Hach YAB019 4x 4-20 mA Output Board for SC1000/SC1500 Controllers and 5500 SC Analyzers
Catalog No. 5000207194 Shop All Hach Company Products
Encompass
The Hach YAB019 is a 4x 4-20 mA output board that provides four isolated analog output channels to expand the analog communication capability of SC1000 and SC1500 controllers and 5500 sc analyzers. It enables external monitoring, control, and integration using standard 4-20 mA signals.
  • Adds four isolated 4-20 mA analog output channels for external systems.
  • Expands analog communication and control capability of compatible controllers and analyzers.
  • Enables integration with external monitoring and control equipment using standard signals.
  • Installs into available accessory slots for straightforward deployment.
  • Provides dedicated analog outputs to improve process monitoring and control.
Catalog No. 50-002-07194 Supplier Hach Company Supplier No. YAB019
The Hach YAB019 is a 4x 4-20 mA output board that provides four isolated analog output channels to expand the analog communication capability of SC1000 and SC1500 controllers and 5500 sc analyzers. It enables external monitoring, control, and integration using standard 4-20 mA signals.
  • Adds four isolated 4-20 mA analog output channels for external systems.
  • Expands analog communication and control capability of compatible controllers and analyzers.
  • Enables integration with external monitoring and control equipment using standard signals.
  • Installs into available accessory slots for straightforward deployment.
  • Provides dedicated analog outputs to improve process monitoring and control.