Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Hack the Planet: Using CRISPR to Terraform Mars Kit
SDPSCI_ED

Catalog No. S27554 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
11 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27554 11 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27554 Supplier Edvotek™ Supplier No. 310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simulate the use of CRISPR-Cas9 to modify bacterial DNA, which will then be transformed into auxotrophic E. coli that are incapable of surviving on the Martian surface. Only bacteria that receive the successfully edited DNA can survive, thrive, and help to terraform Mars!

  • Storage: Some Components Require Freezer Storage Upon Receipt
  • Time Range: Three 1 hr. Modules
  • For 5 Groups

Specifications

Grade 11 to Undergraduate
Age 16 to 20
Includes Instructions, Purified DNA, Simulated sgRNA and Cas9 enzyme, Auxotrophic BactoBeads™, Calcium Chloride, Kanamycin, ReadyPour™ Luria Broth Agar, ReadyPour™ Minimal Media Agar, Minimal Media for Recovery, Growth Media, Assay Components, Plastic Supplies
Requires Incubation Oven, Water Baths, Microwave/Hot Plate, Adjustable Volume Micropipette and Tips, Balance, Centrifuge, Microcentrifuge, Glassware/Cuvettes, Distilled Water, and Ice
Time Range 3, 1 Hr. Modules
Content And Storage Some Components Require Freezer Storage Upon Receipt
Class Size 5 Lab Groups
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.