Learn More
Medchemexpress LLC HADA hydrochloride | 2253733-10-5 | 98.0% | 250 MG
SDP
Catalog No. 5000341779
Encompass_Preferred
HADA hydrochloride is a blue fluorescent D-amino acid that is efficiently incorporated into the peptidoglycans of diverse bacterial species at the sites of peptidoglycan biosynthesis. This allows for specific and covalent probing of bacterial growth with minimal perturbation.
  • Blue fluorescent D-amino acid (λem~450 nm)
  • Efficiently incorporated into peptidoglycans of diverse bacterial species
  • Allows specific and covalent probing of bacterial growth
  • Causes minimal perturbation to bacterial growth
Catalog No. 50-003-41779 Supplier Medchemexpress LLC Supplier No. 250MGHY131045
HADA hydrochloride is a blue fluorescent D-amino acid that is efficiently incorporated into the peptidoglycans of diverse bacterial species at the sites of peptidoglycan biosynthesis. This allows for specific and covalent probing of bacterial growth with minimal perturbation.
  • Blue fluorescent D-amino acid (λem~450 nm)
  • Efficiently incorporated into peptidoglycans of diverse bacterial species
  • Allows specific and covalent probing of bacterial growth
  • Causes minimal perturbation to bacterial growth