Learn More
Medchemexpress LLC HADA hydrochloride | 2253733-10-5 | 98.01% | 100 MG
SDP
Catalog No. 5000340206
Encompass_Preferred
HADA hydrochloride is a blue (λem~450 nm) fluorescent D-amino acid (FDAA). FDAAs are effectively incorporated into the peptidoglycans (PGs) of various bacterial species at PG biosynthesis sites, allowing for specific and covalent probing of bacterial growth with minimal disturbance. For research use only; not sold to patients.
  • Blue fluorescent D-amino acid (FDAA)
  • Efficiently incorporated into peptidoglycans (PGs) of diverse bacterial species
  • Enables specific and covalent probing of bacterial growth
  • Minimal perturbation of bacterial growth
  • For research use only
Catalog No. 50-003-40206 Supplier Medchemexpress LLC Supplier No. HY131045100MG
HADA hydrochloride is a blue (λem~450 nm) fluorescent D-amino acid (FDAA). FDAAs are effectively incorporated into the peptidoglycans (PGs) of various bacterial species at PG biosynthesis sites, allowing for specific and covalent probing of bacterial growth with minimal disturbance. For research use only; not sold to patients.
  • Blue fluorescent D-amino acid (FDAA)
  • Efficiently incorporated into peptidoglycans (PGs) of diverse bacterial species
  • Enables specific and covalent probing of bacterial growth
  • Minimal perturbation of bacterial growth
  • For research use only