Learn More
Aviva Systems Biology Corp HADH Antibody - middle region (ARP54764_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54764_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HADH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human HADH. Alternate names: HAD, HADH1, HADHSC, HHF4, M/SCHAD, MGC8392, SCHAD, HCDH, MSCHADApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-966