Learn More
Aviva Systems Biology Corp HADHB Antibody - C-terminal region (ARP48133_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48133_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HADHB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human HADHB. Alternate names: MGC87480, MSTP029, TP-BETA, ECHB, MTPBApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59722