Learn More
Sino Biological Human mastadenovirus B (HAdV-B) encapsidation protein IVa2 Recombinant (His Tag)
Supplier:  Sino Biological 40630V07B100
Encompass_Preferred
A DNA sequence encoding the Human mastadenovirus B (HAdV-B) encapsidation protein IVa2 (YP_002213830.1) (Thr3-Lys448) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3692