Learn More
New England Biolabs, Inc. Haelll Methyltransferase – 500 units
SDP
Catalog No. 50811646
Encompass
HaeIII Methyltransferase modifies the internal cytosine residue (C5) of the sequence GGCC. This enzyme comes from an E. coli strain that carries the cloned HaeIII modification gene from Haemophilus aegyptius (ATCC 11116).
Catalog No. 50-811-646 Supplier New England Biolabs, Inc. Supplier No. M0224S
HaeIII Methyltransferase modifies the internal cytosine residue (C5) of the sequence GGCC. This enzyme comes from an E. coli strain that carries the cloned HaeIII modification gene from Haemophilus aegyptius (ATCC 11116).