Learn More
New England Biolabs, Inc. HaeII – 10000 units
SDP
Catalog No. 50811936
Encompass
A restriction endonuclease that recognizes the sequence R_GCGC^Y.
Catalog No. 50-811-936 Supplier New England Biolabs, Inc. Supplier No. R0107L
A restriction endonuclease that recognizes the sequence R_GCGC^Y.