Learn More
New England Biolabs, Inc. HaeIII – 3000 units
SDP
Catalog No. 50811940
Encompass
A restriction endonuclease that recognizes the sequence GG_^CC.
Catalog No. 50-811-940 Supplier New England Biolabs, Inc. Supplier No. R0108S
A restriction endonuclease that recognizes the sequence GG_^CC.