Learn More
New England Biolabs, Inc. HaeIII – 15000 units
SDP
Catalog No. 50811939
Encompass
A restriction endonuclease that recognizes the sequence GG_^CC.
Catalog No. 50-811-939 Supplier New England Biolabs, Inc. Supplier No. R0108M
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence GG_^CC.