Learn More
New England Biolabs, Inc. HaeIII – 3000 units
SDP
Supplier:  New England Biolabs, Inc. R0108T
Encompass
A restriction endonuclease that recognizes the sequence GG_^CC.
Catalog No. 50-811-941