Learn More
Hettich Centrifuges HAEMATOKRIT 200, (24) Package
SDP
Catalog No. 502537687 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich HAEMATOKRIT PACKAGE 24, (1) Centrifuge, (1) 24-place Hematocrit Rotor, (1) Lid, Evaluation disc, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7687 Supplier Hettich Centrifuges Supplier No. 200HEMA24
Hettich HAEMATOKRIT PACKAGE 24, (1) Centrifuge, (1) 24-place Hematocrit Rotor, (1) Lid, Evaluation disc, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years