Learn More
Haemo-Sol HAEMO-SOL NON-PHOSPHATE 5LB
SDP
Catalog No. NC0483056
Encompass_Preferred
Haemo-Sol Non-Phosphate (026-058) is specifically formulated to retain all of the advantages of Haemo-Sol Regular, while being especially friendly to the environment. It is particularly effective in soak- and rinse-type situations. It is recommended that customers use � ounce (14 grams) to 1 gallon (3.79 liters) of water as a mix ratio. Haemo-Sol Non-Phosphate contains an anionic surfactant.
Catalog No. NC0483056 Supplier Haemo-Sol Supplier No. 026058
Haemo-Sol Non-Phosphate (026-058) is specifically formulated to retain all of the advantages of Haemo-Sol Regular, while being especially friendly to the environment. It is particularly effective in soak- and rinse-type situations. It is recommended that customers use � ounce (14 grams) to 1 gallon (3.79 liters) of water as a mix ratio. Haemo-Sol Non-Phosphate contains an anionic surfactant.