Learn More
Haemo-Sol Haemo-Sol Regular 6x5lb Must Buy 5
SDP
Catalog No. NC0888464
Encompass_Preferred
Haemo-Sol Regular (026-050) is a great all purpose detergent. It is designed to work in soak- and rinse-type situations, as well as ultrasonic cleaners. Immediate immersion of soiled articles in a Haemo-Sol bath allows for shorter wash cycles. It is recommended that customers use ounce (14 grams) to 1 gallon (3.79 liters) of water as a mix ratio. Haemo-Sol Regular contains an anionic surfactant.
Catalog No. NC0888464 Supplier Haemo-Sol Supplier No. 026050MOQ 5
Haemo-Sol Regular (026-050) is a great all purpose detergent. It is designed to work in soak- and rinse-type situations, as well as ultrasonic cleaners. Immediate immersion of soiled articles in a Haemo-Sol bath allows for shorter wash cycles. It is recommended that customers use ounce (14 grams) to 1 gallon (3.79 liters) of water as a mix ratio. Haemo-Sol Regular contains an anionic surfactant.