Learn More
HTL-Strefa DropSafe Haemolance Plus™ Safety Lance
SDP
Catalog No. 22192014
Change view
Click to view available options
Color:
Blue
Green
Yellow
Depth (Metric) Needle:
1.4 mm
1.8 mm
Needle Gauge:
18 G
21 G
25 G
Flow Type:
High
Low
Normal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Depth (Metric) Needle Needle Gauge Flow Type
22-192-014 Green 1.8 mm 21 G Normal
22-192-015 Yellow 1.8 mm 18 G High
22-192-013 Blue 1.4 mm 25 G Low
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-192-014 Supplier HTL-Strefa Supplier No. 7372
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safety lancets designed for safety, comfort, and simplicity. Fully hidden needle. Silicone-coated, ultra-sharp needle. Easy to handle, intuitive activation. Designed for convenience and ideal blood sample collection.
  • Button Activated Safety lancets designed for safety, comfort, and simplicity.
  • Safety
    • All needles are irradiation-sterilized and protected with a protective cap. Fully enclosed needle housing ensures that the needle is hidden before and after use to prevent sharps injuries. Auto-Retraction mechanism prevents reuse of the device.
  • Comfort
    • Silicone-coated, ultra-sharp needles, ideally positioned for skin penetration. Various, color-coded product versions to meet specific blood sample applications. Wide range of designs providing a simple, comfortable, and secure puncture.
  • Simplicity
    • Easy to handle, intuitive activation with no pre-loading for both push button and pressure activations. Easy to grasp protective cap for easy removal. Simply twist and pull to remove the protective cap. Designed for convenient finger positioning and ideal blood sample collection.
  • Ready to Use
    • The lancet is activated by first removing the safety cap, pressing the lancet against the fingertip, then depressing the push-button.
  • In use
    • During activation, the wings on the needle support break, which releases the needle.
  • After use
    • After activation, the needle retracts back into the device. Immediately dispose of into an appropriate sharps disposal
Specifications
Disposable Single-use
Product Type Lancet
Quantity 100/Box
Depth (Metric) Needle 1.8 mm
Needle Gauge 21 G
Color Green
Flow Type Normal
Sterility Sterile