Learn More
ATCC pfizer Pneumococcal polysaccharide powder Type 12 (d12F) 2mg
Catalog No. 502380116
Encompass
Purified pneumococcal polysaccharide type 12F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 12F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-0116 Supplier ATCC Supplier No. 515X
Purified pneumococcal polysaccharide type 12F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 12F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.