Learn More
Thermo Scientific™ Haemophilus ID Quad Medium

Description
Speciate the genus Haemophilus using Thermo Scientific™ Haemophilus ID Quad, a medium containing three separate growth requirements and horse blood for the production of hemolysis, all on a single prepared media plate.
Read more results from a single sample and produce more diagnostic details while reducing storage capacity issues in the incubator with a quad-plate that includes four separate media formulations.
- One quadrant of the plate contains X factor
- One quadrant contains V factor
- One quadrant contains XV factors
- One quadrant contains XV factors with horse blood
Haemophilus are small, pleomorphic, gram-negative coccobacilli. They are nonmotile, non-sporeforming, fastidious, facultative anaerobes. Almost all species require preformed growth factors present in the blood, particularly the X factor (hemin) and/or V factor (NAD) for growth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Clear agar light to med amber/ clear agar light amber/ clear agar light to med amber/ Opaque red |
| Content And Storage | 2°C to 8°C |
| Description | Haemophilus ID Quad Medium |
| Format | 85mm Quadplate |
| Manufacturing Origin | United States |
| Product Type | Agar |
| Fill Volume | 5 mL |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.