Learn More
ATCC Haemophilus influenzae
Supplier:  ATCC 33391
Encompass
Haemophilus influenzae strain NCTC 8143 is a whole-genome sequenced type strain that has applications in infectious disease research, media testing, and respiratory disease research.
Catalog No. 50-238-4368