Learn More
ATCC Haemophilus influenzae
Catalog No. 502385038
Encompass
Haemophilus influenzae strain AmMS 120 is a whole-genome sequenced clinical isolate that can be used in respiratory disease research or as a quality control strain for MicroScan products.
Catalog No. 50-238-5038 Supplier ATCC Supplier No. 49144
Haemophilus influenzae strain AmMS 120 is a whole-genome sequenced clinical isolate that can be used in respiratory disease research or as a quality control strain for MicroScan products.