Learn More
ATCC Haemophilus influenzae
Catalog No. 502384255
Encompass
Haemophilus influenzae strain L-378 was isolated from a lung abscess. This item is a quality control strain for Sensititre products and for CLSI antibiotic disk susceptibility testing.
Catalog No. 50-238-4255 Supplier ATCC Supplier No. 49766
Haemophilus influenzae strain L-378 was isolated from a lung abscess. This item is a quality control strain for Sensititre products and for CLSI antibiotic disk susceptibility testing.