Learn More
ATCC Haemophilus influenzae
Catalog No. 502384194
Encompass
Haemophilus influenzae strain TD-4 was isolated in January 1984 from the expectorated sputum of a male with pneumonia. This whole-genome sequenced bacterial strain can be used in susceptibility disc testing and respiratory disease research.
Catalog No. 50-238-4194 Supplier ATCC Supplier No. 49247
Haemophilus influenzae strain TD-4 was isolated in January 1984 from the expectorated sputum of a male with pneumonia. This whole-genome sequenced bacterial strain can be used in susceptibility disc testing and respiratory disease research.