Learn More
ATCC Rhizobium tropici
Supplier:  ATCC 49672
Encompass
Rhizobium tropici strain CIAT 899 is a bacterial type strain that was isolated in 1978 from nodules on Phaseolus vulgaris in Colombia. This strain is propagated aerobically in Rhizobium X medium.
Catalog No. 50-238-1308