Learn More
ATCC Haemophilus influenzae; Strain KW20; genomic DNA
Catalog No. 502381844
Encompass
Genomic DNA is isolated from Haemophilus influenzae strain Rd (ATCC 51907). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-1844 Supplier ATCC Supplier No. 51907D5
Genomic DNA is isolated from Haemophilus influenzae strain Rd (ATCC 51907). This whole-genome sequenced product can be used in PCR and other molecular biology applications.