Learn More
ATCC Haemophilus parahaemolyticus; 536
Catalog No. 502383069
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Haemophilus parahaemolyticus strain 536 [NCTC 8479] was isolated from a chronic pulmonary infection. This bacterial type strain is known to produce the HhaI and HhaII restriction endonucleases.
Catalog No. 50-238-3069 Supplier ATCC Supplier No. 10014
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Haemophilus parahaemolyticus strain 536 [NCTC 8479] was isolated from a chronic pulmonary infection. This bacterial type strain is known to produce the HhaI and HhaII restriction endonucleases.