Learn More
ATCC Quantitative Genomic DNA from; Haemophilus ducreyi; Strain: CIP 542
Supplier:  ATCC 33940DQ
Encompass
Quantitative genomic DNA from Haemophilus ducreyi strain CIP 542 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-1252