Learn More
ATCC Yersinia enterocolitica; subsp. enterocolitica; Strain WA-314
Catalog No. 502379581
Encompass
Depositor states that this strain carries the 75 kb virulence plasmid pYV08.
Catalog No. 50-237-9581 Supplier ATCC Supplier No. 51871
Depositor states that this strain carries the 75 kb virulence plasmid pYV08.