Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium boride, 99.5% (metals basis excluding Zr), Zr <2%, Thermo Scientific Chemicals

Catalog No. AA4366430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4366430 250 g
AA4366409 10 g
AA4366418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4366430 Supplier Thermo Scientific Chemicals Supplier No. 04366430
Add to Cart

missing translation for 'validMainframeMsg'

Hafnium boride is used in wear resistant coatings. It is also employed as a nuclear reactor control rod. Furthermore, it is used in hypervelocity re-entry vehicles like ICBM heat shields or aerodynamic leading-edges. It is useful in the preparation of high temperature composites in conjunction with silicon carbide (SiC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-23-7
Molecular Formula B2Hf
MDL Number MFCD00016125

Specifications

Melting Point 3250°C
Physical Form Powder
Quantity 250 g
Linear Formula HfB2
Solubility Information Insoluble in water.
Formula Weight 200.11
Percent Purity 99.5%
Odor Odorless
Density 10.5 g/mL
Assay (metals basis excluding Zr), Zr <2%
Chemical Name or Material Hafnium boride
Show More Show Less
EINECSNumber 234-500-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.