Learn More
Strem, An Ascensus Company CAS# 13499-05-3. 10g. Hafnium(IV) chloride, sublimed grade (99.9+%-Hf, <1.0% Zr). MFCD00011034
SDP
Supplier:  Strem, An Ascensus Company 72075110G
Encompass
CAS# 13499-05-3. 10g. Hafnium(IV) chloride, sublimed grade (99.9+%-Hf, <1.0% Zr). MFCD00011034. Molecular Weight: 320.30. Molecular Formula: HfCl4. Color/form: white pwdr. Strem# 72-0751. http://www.strem.com/catalog/v/72-0751/
Catalog No. 50-901-12927