Learn More
Strem, An Ascensus Company CAS# 13499-05-3. 5g. Hafnium(IV) chloride, sublimed grade (99.9+%-Hf, <0.05% Zr). MFCD00011034
SDP
Catalog No. 5090112931
Encompass
CAS# 13499-05-3. 5g. Hafnium(IV) chloride, sublimed grade (99.9+%-Hf, <0.05% Zr). MFCD00011034. Molecular Weight: 320.30. Molecular Formula: HfCl4. Color/form: white pwdr. Strem# 72-0752. http://www.strem.com/catalog/v/72-0752/
Catalog No. 50-901-12931 Supplier Strem, An Ascensus Company Supplier No. 7207525G
CAS# 13499-05-3. 5g. Hafnium(IV) chloride, sublimed grade (99.9+%-Hf, <0.05% Zr). MFCD00011034. Molecular Weight: 320.30. Molecular Formula: HfCl4. Color/form: white pwdr. Strem# 72-0752. http://www.strem.com/catalog/v/72-0752/