Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium(IV) isopropoxide isopropanol adduct, 99%

Catalog No. AAH5228814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5228814 25 g
AAH5228822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5228814 Supplier Thermo Scientific Chemicals Supplier No. H5228814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2171-99-5 | C12H32HfO4 | 418.874 g/mol

Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2171-99-5
Molecular Formula C12H32HfO4
Molecular Weight (g/mol) 418.874
MDL Number MFCD00070460
InChI Key HMKGKDSPHSNMTM-UHFFFAOYSA-N
Synonym propan-2-ol-hafnium 4/1, 2-propanol, hafnium 4+ salt 9ci
PubChem CID 74764872
IUPAC Name hafnium;propan-2-ol
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Hf]

Specifications

Linear Formula Hf(OOCH(CH3)2)4·HOCH(CH3)2
Quantity 25 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 474.94
Percent Purity 99%
Chemical Name or Material Hafnium(IV) isopropoxide isopropanol adduct
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.