Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium(IV) oxide, 99.95%, (metals basis excluding Zr), Zr typically <0.5%, Thermo Scientific Chemicals

Catalog No. AA3566614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3566614 25 g
AA3566603 1 g
AA3566606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3566614 Supplier Thermo Scientific Chemicals Supplier No. 03566614
Add to Cart

missing translation for 'validMainframeMsg'

Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. It is also used in the preparation of hafnium tetrachloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12055-23-1
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Melting Point 2774°C
Physical Form Powder
Quantity 25 g
Merck Index 14,4588
Solubility Information Insoluble in water.
Formula Weight 210.49
Percent Purity 99.95%
Odor Odorless
Density 9.68 g/mL
Assay (metals basis excluding Zr), Zr typically <0.5%
Chemical Name or Material Hafnium(IV) oxide
Show More Show Less
EINECSNumber 235-013-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.