Learn More
Strem, An Ascensus Company CAS# 7440-58-6. 10g. Hafnium powder (99.6%, Zr-2.2%). MFCD00011032
SDP
Catalog No. 5090116265
Encompass
CAS# 7440-58-6. 10g. Hafnium powder (99.6%, Zr-2.2%). MFCD00011032. Molecular Weight: 178.49. Molecular Formula: Hf. Color/form: -325 mesh Strem# 93-7227. http://www.strem.com/catalog/v/93-7227/
Catalog No. 50-901-16265 Supplier Strem, An Ascensus Company Supplier No. 93722710G
CAS# 7440-58-6. 10g. Hafnium powder (99.6%, Zr-2.2%). MFCD00011032. Molecular Weight: 178.49. Molecular Formula: Hf. Color/form: -325 mesh Strem# 93-7227. http://www.strem.com/catalog/v/93-7227/