Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnocene Dichloride 98.0+%, TCI America™
SDP

Catalog No. H09141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09141G 1 g
H09145G 5 g
H091425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H09141G Supplier TCI America Supplier No. H09141G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12116-66-4
Molecular Formula C10H10Cl2Hf
Molecular Weight (g/mol) 379.58
MDL Number MFCD00001438,MFCD00001438
InChI Key CSEGCHWAMVIXSA-UHFFFAOYSA-L
Synonym bis cyclopentadienyl hafniumdichloride
PubChem CID 131674741
IUPAC Name hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride
SMILES [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1

Specifications

Melting Point 233°C
Quantity 1 g
Formula Weight 379.58
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hafnocene Dichloride
RTECSNumber MG4815000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.