Learn More
Grainger Action Chemical Hairnet, Nylon, Blue, 24 in., 1,000/Pk.
Catalog No. 502771393
Encompass_Preferred
These nylon hairnets are designed for use in professional environments requiring contamination control and hair restraint. Featuring a 24 in. diameter and a honeycomb netting style, they provide a secure fit for comfortable, long-term wear.
  • Color: Blue
  • Material: Nylon
  • Diameter: 24 in.
  • Honeycomb netting style
  • Lint-free
  • Latex-free
  • Package quantity: 1,000 per box
Catalog No. 50-277-1393 Supplier Grainger Supplier No. 13V898
These nylon hairnets are designed for use in professional environments requiring contamination control and hair restraint. Featuring a 24 in. diameter and a honeycomb netting style, they provide a secure fit for comfortable, long-term wear.
  • Color: Blue
  • Material: Nylon
  • Diameter: 24 in.
  • Honeycomb netting style
  • Lint-free
  • Latex-free
  • Package quantity: 1,000 per box