Learn More
Medchemexpress LLC HAIYPRH hydrochloride | 100.0% | 929.46 | 5 MG
SDP
Catalog No. 5000423736
Encompass_Preferred
HAIYPRH hydrochloride is a targeting ligand that specifically binds to the transferrin receptor (TfR). It can mediate the transport of nanocarriers across the blood-brain barrier.
  • Can be conjugated to liposomes for ischemic stroke targeting treatment.
  • Heptapeptide that can be used to target gliomas.
  • Used as a ligand to target co-delivery systems to tumor cells expressing transferrin receptors.
  • Modified co-delivery systems show higher efficiency in cellular uptake and gene expression in U87 MG cells.
  • Modified co-delivery systems accumulate in tumors more efficiently in vivo.
Catalog No. 50-004-23736 Supplier Medchemexpress LLC Supplier No. HYP23145MG
HAIYPRH hydrochloride is a targeting ligand that specifically binds to the transferrin receptor (TfR). It can mediate the transport of nanocarriers across the blood-brain barrier.
  • Can be conjugated to liposomes for ischemic stroke targeting treatment.
  • Heptapeptide that can be used to target gliomas.
  • Used as a ligand to target co-delivery systems to tumor cells expressing transferrin receptors.
  • Modified co-delivery systems show higher efficiency in cellular uptake and gene expression in U87 MG cells.
  • Modified co-delivery systems accumulate in tumors more efficiently in vivo.