Learn More
Medchemexpress LLC Haiyprh hydrochloride | 268215-17-4 | 100.0% | 929.46 | 25 MG
SDP
Catalog No. 5000425481
Encompass_Preferred
HAIYPRH hydrochloride is a targeting ligand that specifically binds to the transferrin receptor (TfR) and can facilitate the transport of nanocarriers across the blood-brain barrier. It is also a heptapeptide used to target gliomas and is intended for research use only.
  • Specifically binds to the transferrin receptor (TfR)
  • Facilitates transport of nanocarriers across the blood-brain barrier
  • Used to target gliomas
  • Functions as a ligand for co-delivery systems to tumor cells expressing transferrin receptors
  • Demonstrates higher efficiency in cellular uptake and gene expression in U87 MG cells
  • Accumulates more efficiently in tumors in vivo
  • Appears as a white to off-white solid
  • Soluble in H2O at concentrations of 100 mg/mL or greater
Catalog No. 50-004-25481 Supplier Medchemexpress LLC Supplier No. 25MGHYP2314
HAIYPRH hydrochloride is a targeting ligand that specifically binds to the transferrin receptor (TfR) and can facilitate the transport of nanocarriers across the blood-brain barrier. It is also a heptapeptide used to target gliomas and is intended for research use only.
  • Specifically binds to the transferrin receptor (TfR)
  • Facilitates transport of nanocarriers across the blood-brain barrier
  • Used to target gliomas
  • Functions as a ligand for co-delivery systems to tumor cells expressing transferrin receptors
  • Demonstrates higher efficiency in cellular uptake and gene expression in U87 MG cells
  • Accumulates more efficiently in tumors in vivo
  • Appears as a white to off-white solid
  • Soluble in H2O at concentrations of 100 mg/mL or greater