Learn More
Apexbio Technology LLC HALAZONE-250MG
SDP
Catalog No. 5000570261
Encompass_Preferred
Halazone (CAS No 80-13-7) is a chemical compound studied in the context of antimicrobial research Originating from efforts to develop water purification methods Halazone functions by releasing active chlorinated species targeting pathogenic microorganisms by disrupting cellular metabolism and integrity Its mechanism of action involves oxidation and chlorination pathways proving effective in reducing microbial viability in various conditions In vitro activities are often observed in the nanomolar to low micromolar range indicating potent efficacy in controlled environments Typical applications for Halazone include studies using bacterial cultures and animal models to explore its potential as a disinfectant and therapeutic agent It is also utilized in drug screening to identify compounds with similar antimicrobial properties Experimental concentrations typically depend on the specific research design allowing for tailored investigation into its efficacy and safety profile
Catalog No. 50-005-70261 Supplier Apexbio Technology LLC Supplier No. BA1377250
Halazone (CAS No 80-13-7) is a chemical compound studied in the context of antimicrobial research Originating from efforts to develop water purification methods Halazone functions by releasing active chlorinated species targeting pathogenic microorganisms by disrupting cellular metabolism and integrity Its mechanism of action involves oxidation and chlorination pathways proving effective in reducing microbial viability in various conditions In vitro activities are often observed in the nanomolar to low micromolar range indicating potent efficacy in controlled environments Typical applications for Halazone include studies using bacterial cultures and animal models to explore its potential as a disinfectant and therapeutic agent It is also utilized in drug screening to identify compounds with similar antimicrobial properties Experimental concentrations typically depend on the specific research design allowing for tailored investigation into its efficacy and safety profile