Learn More
Medchemexpress LLC Halazone | 80-13-7 | 75.0% | 250 MG
SDP
Catalog No. 5000414252
Encompass_Preferred
Halazone is an atypical antimicrobial sulfonamide derivative and a carbonic anhydrase II inhibitor with a Kd value of 1.45 μM. It protects sodium channels from inactivation and is widely utilized for the disinfection of drinking water. Chemically, it is closely related to Chloramine-T, differing in that its nitrogen atom is linked to two chlorine atoms instead of one, and a methyl group is replaced by a carboxyl group.
  • Acts as a carbonic anhydrase II inhibitor (Kd value of 1.45 μM).
  • Protects sodium channels from inactivation.
  • Widely used for disinfection of drinking water.
  • Unstable in solutions, requires freshly prepared solutions.
  • Intended for research use only.
Catalog No. 50-004-14252 Supplier Medchemexpress LLC Supplier No. 250MGHYB1386
Halazone is an atypical antimicrobial sulfonamide derivative and a carbonic anhydrase II inhibitor with a Kd value of 1.45 μM. It protects sodium channels from inactivation and is widely utilized for the disinfection of drinking water. Chemically, it is closely related to Chloramine-T, differing in that its nitrogen atom is linked to two chlorine atoms instead of one, and a methyl group is replaced by a carboxyl group.
  • Acts as a carbonic anhydrase II inhibitor (Kd value of 1.45 μM).
  • Protects sodium channels from inactivation.
  • Widely used for disinfection of drinking water.
  • Unstable in solutions, requires freshly prepared solutions.
  • Intended for research use only.