Learn More
Apexbio Technology LLC Halcinonide 3093-35-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062488
Encompass_Preferred
Halcinonide (CAS 3093-35-4) is a potent synthetic corticosteroid widely utilized in biomedical research as a topical anti-inflammatory agent Mechanistically halcinonide exerts biological effects primarily through activation of glucocorticoid receptor pathways subsequently modulating gene transcription and suppressing the release of inflammatory mediators Owing to its robust corticosteroid receptor agonism and pronounced anti-inflammatory properties this compound frequently supports investigations into inflammatory dermatological conditions and glucocorticoid signaling pathways
Catalog No. 50-000-62488 Supplier Apexbio Technology LLC Supplier No. B17665.1
Halcinonide (CAS 3093-35-4) is a potent synthetic corticosteroid widely utilized in biomedical research as a topical anti-inflammatory agent Mechanistically halcinonide exerts biological effects primarily through activation of glucocorticoid receptor pathways subsequently modulating gene transcription and suppressing the release of inflammatory mediators Owing to its robust corticosteroid receptor agonism and pronounced anti-inflammatory properties this compound frequently supports investigations into inflammatory dermatological conditions and glucocorticoid signaling pathways